KEY465194591 Key Organics / BIONET

3-(2-methylbenzenesulfonyl)quinolin-2-amine

  • MDL number MFCD03848438
  • Cas Number : 861210-75-5
  • Molecular Weight : 298.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69791

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2W-0875_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart