KEY465194596 Key Organics / BIONET

1,3-diethyl 2-{[(2,3-dimethylquinoxalin-6-yl)amino]methylidene}propanedioate

  • MDL number MFCD03618048
  • Cas Number : 170948-50-2
  • Molecular Weight : 343.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69811

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2W-0902_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart