KEY465194597 Key Organics / BIONET

1-(2,3-dimethylquinoxalin-6-yl)-3-(prop-2-en-1-yl)thiourea

  • MDL number MFCD03618049
  • Cas Number : 861210-91-5
  • Molecular Weight : 272.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69812

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2W-0903_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart