KEY465194601 Key Organics / BIONET

2-[2-(pyridin-2-yl)-1,3-thiazol-4-yl]ethan-1-amine hydrobromide

  • MDL number MFCD04124954
  • Cas Number : 1052552-07-4
  • Molecular Weight : 286.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69821

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2X-0208_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart