KEY465194606 Key Organics / BIONET

4-({2-[2-(4-methylphenyl)-1,3-thiazol-4-yl]ethyl}amino)-3-nitrobenzoic acid

  • MDL number MFCD04124968
  • Cas Number : 861211-10-1
  • Molecular Weight : 383.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69840

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2X-0259_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart