KEY465194613 Key Organics / BIONET

[(E)-[(2,5-dimethoxyphenyl)methylidene]amino]thiourea

  • MDL number MFCD00449245
  • Cas Number : 1254353-86-0
  • Molecular Weight : 239.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69857

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2X-0293_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart