KEY465194617 Key Organics / BIONET

ethyl 2-{[4-(3-fluoro-4-methoxyphenyl)-5-methylpyrimidin-2-yl]amino}-1,3-thiazole-4-carboxylate

  • MDL number MFCD05668751
  • Cas Number : 861211-24-7
  • Molecular Weight : 388.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69862

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2X-0302_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart