KEY465194618 Key Organics / BIONET

methyl 6-amino-5-cyano-2-(2-methoxy-2-oxoethyl)-4-(4-methylphenyl)-4H-pyran-3-carboxylate

  • MDL number MFCD04124987
  • Cas Number : 547769-52-8
  • Molecular Weight : 342.351

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69866

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2X-0311_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart