KEY465194619 Key Organics / BIONET

(6-oxo-5-propanoyl-1,6-dihydropyrimidin-2-yl)thiourea

  • MDL number MFCD04124989
  • Cas Number : 861211-26-9
  • Molecular Weight : 226.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69868

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2X-0315_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart