KEY465194620 Key Organics / BIONET

[6-oxo-4-(trifluoromethyl)-1,6-dihydropyrimidin-2-yl]thiourea

  • MDL number MFCD04124990
  • Cas Number : 861211-27-0
  • Molecular Weight : 238.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69869

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2X-0318_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart