KEY465194622 Key Organics / BIONET

5-[(4-methylphenyl)methyl]-4-oxo-1H,2H,3H,3aH,4H,5H-pyrrolo[1,2-a]quinoxaline-7-carbonitrile

  • MDL number MFCD04124992
  • Cas Number : 1008710-16-4
  • Molecular Weight : 317.392

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69871

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2X-0321_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart