KEY465194624 Key Organics / BIONET

(E)-N'-(5-{[1,1'-biphenyl]-4-carbonyl}-1,3-thiazol-2-yl)-N,N-dimethylmethanimidamide

  • MDL number MFCD04124995
  • Cas Number : 861211-31-6
  • Molecular Weight : 335.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69874

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2X-0328_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart