KEY465194628 Key Organics / BIONET

(2E)-2-[4-(2-fluorophenyl)-1,3-thiazol-2-yl]-3-(4-oxo-4H-chromen-3-yl)prop-2-enenitrile

  • MDL number MFCD04125009
  • Cas Number : 861211-44-1
  • Molecular Weight : 374.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69910

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2X-0822_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart