KEY465194630 Key Organics / BIONET

3-{[(6-chloropyridin-3-yl)methyl](methyl)amino}propanenitrile

  • MDL number MFCD04125025
  • Cas Number : 861211-56-5
  • Molecular Weight : 209.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69931

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2X-0853_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart