KEY465194633 Key Organics / BIONET

2-[4-(4-fluorobenzoyl)-1H-1,2,3-triazol-1-yl]-4-methyl-6-(trifluoromethyl)pyrimidine

  • MDL number MFCD05668758
  • Cas Number : 861211-62-3
  • Molecular Weight : 351.265

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69938

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/2X-0867_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart