KEY465194637 Key Organics / BIONET

2-{6-oxo-5,12,14-trioxa-2-azatetracyclo[7.7.0.0^{3,7}.0^{11,15}]hexadeca-1(9),3(7),10,15-tetraen-8-yl}benzoic acid

  • MDL number MFCD08064720
  • Cas Number : 882747-22-0
  • Molecular Weight : 351.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69954

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2Y-0005_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart