KEY465194640 Key Organics / BIONET

8-(2,4-dichlorophenyl)-5,12,14-trioxa-2-azatetracyclo[7.7.0.0^{3,7}.0^{11,15}]hexadeca-1(9),3(7),10,15-tetraen-6-one

  • MDL number MFCD08064722
  • Cas Number : 882747-24-2
  • Molecular Weight : 376.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69958

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2Y-0092_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart