KEY465194646 Key Organics / BIONET

1,3-dimethyl 2-[2-nitro-5-(trifluoromethyl)phenyl]-2-(prop-2-yn-1-yl)propanedioate

  • MDL number MFCD07021356
  • Cas Number : 900019-64-9
  • Molecular Weight : 359.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69965

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2Y-0256_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart