KEY465194648 Key Organics / BIONET

ethyl 4-[(1-oxo-2,3-dihydro-1H-isoindol-2-yl)methyl]benzoate

  • MDL number MFCD06496169
  • Cas Number : 861211-67-8
  • Molecular Weight : 295.338

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69972

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2Y-0800_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart