KEY465194654 Key Organics / BIONET

N-[2-(1H-indol-3-yl)ethyl]-3,4-dimethoxybenzene-1-sulfonamide

  • MDL number MFCD08064729
  • Cas Number : 880139-08-2
  • Molecular Weight : 360.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69985

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2Y-0816_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart