KEY465194660 Key Organics / BIONET

(2Z)-2-[(Z)-3-fluoro-4-(morpholin-4-yl)benzoyl]-3-(3-methoxyphenyl)prop-2-enenitrile

  • MDL number MFCD07021363
  • Cas Number : 900019-69-4
  • Molecular Weight : 366.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70001

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2Y-0834_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart