KEY465194664 Key Organics / BIONET

2-amino-7-bromo-4-(pyridin-3-yl)-4H-benzo[g]chromene-3-carbonitrile

  • MDL number MFCD08064732
  • Cas Number : 882747-31-1
  • Molecular Weight : 378.229

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70011

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2Y-0929_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart