KEY465194674 Key Organics / BIONET

ethyl 3,5-dimethyl-1-[3-(trifluoromethyl)phenyl]-1H-pyrazole-4-carboxylate

  • MDL number MFCD07021370
  • Cas Number : 956950-76-8
  • Molecular Weight : 312.292

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70066

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/2Z-0811_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart