KEY465194683 Key Organics / BIONET

(4Z)-1-(4,5-dihydro-1,3-thiazol-2-yl)-3-methyl-4-{2-[3-(trifluoromethyl)phenyl]hydrazin-1-ylidene}-4,5-dihydro-1H-pyrazol-5-one

  • MDL number MFCD00140899
  • Cas Number : 338402-56-5
  • Molecular Weight : 355.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70115

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3B-062_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart