KEY465194687 Key Organics / BIONET

N-[(E)-2-(3-methyl-4-nitro-1,2-oxazol-5-yl)ethenyl]-4-(trifluoromethoxy)aniline

  • MDL number MFCD00127355
  • Cas Number : 338402-61-2
  • Molecular Weight : 329.235

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70129

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3C-003_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart