KEY465194689 Key Organics / BIONET

N-[(E)-2-(1,3-dimethyl-4-nitro-1H-pyrazol-5-yl)ethenyl]-4-(trifluoromethoxy)aniline

  • MDL number MFCD00214727
  • Cas Number : 436100-82-2
  • Molecular Weight : 342.278

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70140

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3C-018_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart