KEY465194705 Key Organics / BIONET

(2E)-2-(4-chlorophenyl)-3-[(2-methylphenyl)amino]prop-2-enenitrile

  • MDL number MFCD11656734
  • Cas Number : 338403-04-6
  • Molecular Weight : 268.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70166

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3C-104_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart