KEY465194709 Key Organics / BIONET

4-{[(E)-2-[3-(2-chloro-6-fluorophenyl)-4-cyano-1,2-oxazol-5-yl]ethenyl]amino}benzoic acid

  • MDL number MFCD00127381
  • Cas Number : 338403-15-9
  • Molecular Weight : 383.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70174

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3C-115_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart