KEY465194714 Key Organics / BIONET

4-[(2E)-3-(dimethylamino)-2-phenylprop-2-enoyl]-N,N-dimethyl-1H-pyrrole-2-carboxamide

  • MDL number MFCD00141862
  • Cas Number : 338403-38-6
  • Molecular Weight : 311.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70192

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3D-019_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart