KEY465194720 Key Organics / BIONET

(2E)-3-[4-(2-chlorophenoxy)-3-nitrophenyl]prop-2-enoic acid

  • MDL number MFCD00138791
  • Cas Number : 338403-65-9
  • Molecular Weight : 319.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70214

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/3D-058_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart