KEY465194763 Key Organics / BIONET

N-(5-acetyl-6-methyl-2-oxo-2H-pyran-3-yl)-4-methoxybenzamide

  • MDL number MFCD00139788
  • Cas Number : 338404-98-1
  • Molecular Weight : 301.298

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70404

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3E-945_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart