KEY465194769 Key Organics / BIONET

4-chloro-1-(4-chlorophenyl)-6-oxo-1,6-dihydropyridazine-3-carbonitrile

  • MDL number MFCD00141174
  • Cas Number : 338405-10-0
  • Molecular Weight : 266.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70417

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3F-012_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart