KEY465194775 Key Organics / BIONET

1-{N'-[2-(1H-1,2,4-triazol-1-yl)ethanimidoyl]hydrazinecarbonyl}-N-[4-(trifluoromethoxy)phenyl]formamide

  • MDL number MFCD00214731
  • Cas Number : 338405-27-9
  • Molecular Weight : 371.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70434

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3F-038_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart