KEY465194776 Key Organics / BIONET

2-[(1-phenyl-1H-1,2,4-triazol-3-yl)oxy]-N-(phenylamino)ethanimidamide

  • MDL number MFCD00214733
  • Cas Number : 338405-30-4
  • Molecular Weight : 308.345

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70437

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3F-041_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart