KEY465194782 Key Organics / BIONET

N'-[2-(2,4-dichlorophenoxy)ethanimidoyl]-1H-pyrrole-2-carbohydrazide

  • MDL number MFCD01936205
  • Cas Number : 400080-24-2
  • Molecular Weight : 327.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70459

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3F-068_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart