KEY465194784 Key Organics / BIONET

N-phenyl-1-{N'-[2-(1H-1,2,4-triazol-1-yl)ethanimidoyl]hydrazinecarbonyl}formamide

  • MDL number MFCD00172031
  • Cas Number : 338405-44-0
  • Molecular Weight : 287.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70461

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3F-070_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart