KEY465194795 Key Organics / BIONET

methyl (2Z)-3-amino-4-(1H-1,2,4-triazol-1-yl)but-2-enoate

  • MDL number MFCD00214739
  • Cas Number : 338405-78-0
  • Molecular Weight : 182.183

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70488

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3F-100_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart