KEY465194802 Key Organics / BIONET

methyl 2-({4-[(1E)-(methoxyimino)methyl]-1-methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl}sulfanyl)benzoate

  • MDL number MFCD00140864
  • Cas Number : 321553-57-5
  • Molecular Weight : 373.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70574

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3F-405S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart