KEY465194805 Key Organics / BIONET

ethyl 5-(3,4-dichlorophenyl)-2-(morpholin-4-yl)-1H-pyrrole-3-carboxylate

  • MDL number MFCD00171800
  • Cas Number : 338406-26-1
  • Molecular Weight : 369.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70595

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3F-922_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart