KEY465194815 Key Organics / BIONET

2-(5-benzoyl-1,2-oxazol-3-yl)-3-chloro-5-(trifluoromethyl)pyridine

  • MDL number MFCD01315022
  • Cas Number : 478030-83-0
  • Molecular Weight : 352.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70637

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3G-021_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart