KEY465194823 Key Organics / BIONET

(2E)-1-(5-bromo-6-methoxynaphthalen-2-yl)-3-(dimethylamino)-2-methylprop-2-en-1-one

  • MDL number MFCD00173451
  • Cas Number : 338407-63-9
  • Molecular Weight : 348.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70679

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3G-081_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart