KEY465194831 Key Organics / BIONET

6-nitro-N-(propan-2-yl)-1,2-benzoxazole-3-carboxamide

  • MDL number MFCD00665688
  • Cas Number : 400080-49-1
  • Molecular Weight : 249.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70702

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3G-318S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart