KEY465194832 Key Organics / BIONET

N-(4-methylphenyl)-3-nitro-4-(1H-1,2,4-triazol-1-yl)benzamide

  • MDL number MFCD00142000
  • Cas Number : 338407-90-2
  • Molecular Weight : 323.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70706

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3G-323S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart