KEY465194846 Key Organics / BIONET

methyl 3-hydroxy-3-(trifluoromethyl)-hexahydro-2H-[1,2]oxazolo[2,3-a]pyridine-2-carboxylate

  • MDL number MFCD00172848
  • Cas Number : 317833-14-0
  • Molecular Weight : 269.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70808

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3G-911_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart