KEY465194851 Key Organics / BIONET

3-chloro-N-{2-[(Z)-{[4-(dimethylamino)phenyl]methylidene}amino]ethyl}-5-(trifluoromethyl)pyridin-2-amine

  • MDL number MFCD00172564
  • Cas Number : 338409-13-5
  • Molecular Weight : 370.80

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70836

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3G-941_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart