KEY465194859 Key Organics / BIONET

3-chloro-N-(3-methoxypropyl)-5-(trifluoromethyl)pyridin-2-amine

  • MDL number MFCD00172036
  • Cas Number : 338406-33-0
  • Molecular Weight : 268.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70867

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3G-975_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart