KEY465194862 Key Organics / BIONET

3-chloro-N-(2,2-dimethoxyethyl)-5-(trifluoromethyl)pyridin-2-amine

  • MDL number MFCD00172038
  • Cas Number : 338406-44-3
  • Molecular Weight : 284.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70872

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3G-983_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart