KEY465194864 Key Organics / BIONET

(E)-N'-({1-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-1H-pyrrol-2-yl}sulfonyl)-N,N-dimethylmethanimidamide

  • MDL number MFCD00214871
  • Cas Number : 338406-54-5
  • Molecular Weight : 380.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70882

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3H-001_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart