KEY465194868 Key Organics / BIONET

1-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-5-nitro-1H-indazole

  • MDL number MFCD00214886
  • Cas Number : 150149-16-9
  • Molecular Weight : 342.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70907

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3H-028_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart