KEY465194873 Key Organics / BIONET

N-[(E)-2-(benzenesulfonyl)-2-nitroethenyl]-2-chloroaniline

  • MDL number MFCD00214898
  • Cas Number : 1164476-20-3
  • Molecular Weight : 338.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70938

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3H-072_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart